Organic reference standard, Droperidol, Ph.Eur., RS
EDQMD3100000
: EDQM
:
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:548-73-2
- Description:Droperidol
- Formula:C₂₂H₂₂FN₃O₂
- Storage temperature:Ambient
- Description:Droperidol
- In Monographs 1008, 1009, 1010, and 1172
- Soluble in/miscible with chloroform, DMF, methanol, and acetone