Organic reference standard, doxycycline hyclate, Ph.Eur., RS
Catalog # EDQMD3000000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:24390-14-5
- Description:Doxycycline hyclate
- Formula:C₄₆H₅₈Cl₂N₄O₁₈
- Storage temperature:Ambient
- Description:Doxycycline hyclate
Specifications
About this item
- In Monographs 0272 and 0820
- Solubility in water: 100 to 1000 g/l