Organic reference standard, Doxorubicin hydrochloride, Ph.Eur., RS
Catalog # EDQMD2975000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:25316-40-9
- Description:Doxorubicin hydrochloride
- Formula:C₂₇H₃₀ClNO₁₁
- Storage temperature:-20°C ± 5°C
- Description:Doxorubicin hydrochloride
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monographs 0714, 0662, and 1590
- Solubility in water: 33,3 - 100 g/l