$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Standards
Organic Standards
Organic reference standards, Domperidone, Ph.Eur., RS
Organic reference standards, Domperidone, Ph.Eur., RS
:
EDQM
Organic reference standards, Domperidone, Ph.Eur., RS
:
EDQM
Domperidone in Monograph 1009. Domperidone maleate in Monograph 1008
CAS No.:
83898-65-1
Formula:
C₂₆H₂₈ClN₅O₆
Storage temperature:
Ambient
Description:
Domperidone maleate