Organic reference standard, Dihydrostreptomycin sulfate, Ph.Eur., RS
Catalog # EDQMD1954000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Reference standard
- CAS No.:5490-27-7
- Description:Dihydrostreptomycin sulfate
- Formula:C₄₂H₈₈N₁₄O₃₆S₃
- Storage temperature:Ambient
- Description:Dihydrostreptomycin sulfate
Specifications
About this item
- In Monograph 0485
- Pharmaceutical industry applications