Organic reference standard, Dihydroergocristine mesilate, Ph.Eur., RS
EDQMD1945000
: EDQM
:
- Pk:250 mg
- Standards:Reference standard
- CAS No.:24730-10-7
- Description:Dihydroergocristine mesilate
- Formula:C₃₆H₄₅N₅O₈S
- Storage temperature:Ambient
- Description:Dihydroergocristine mesilate
- Solubility in water: 0,01 g/l 20 °C
- Pharmaceutical industry applications