Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standard, Dicycloverine hydrochloride, Ph.Eur., RS
  EDQMD1060000
 :  EDQM
 :  
Organic reference standard, Dicycloverine hydrochloride, Ph.Eur., RS
  EDQMD1060000
 :  EDQM
 :  

 

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    67-92-5
  • Description:
    Dicycloverine hydrochloride
  • Formula:
    C₁₉H₃₆CINO₂
  • Storage temperature:
    Ambient
  • Description:
    Dicycloverine hydrochloride

 

 

  • In Monograph 1197
  • Soluble in water