Organic reference standard, Demeclocycline hydrochloride, Ph.Eur., RS
EDQMD0300000
: EDQM
:
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:64-73-3
- Description:Demeclocycline hydrochloride
- Formula:C₂₁H₂₂Cl₂N₂O₈
- Storage temperature:Ambient
- Description:Demeclocycline hydrochloride
- In Monographs 0211, 0210, 0176, 0173, and 1654
- Pharmaceutical industry applications