Organic reference standard, Cyproterone acetate, Ph.Eur., RS
EDQMC3283000
: EDQM
:
- Pack type:Vial
- Pk:30 mg
- Standards:Reference standard
- CAS No.:427-51-0
- Description:Cyproterone acetate
- Formula:C₂₄H₂₉ClO₄
- Storage temperature:Ambient
- Description:Cyproterone acetate
- In Monograph 1094
- Soluble in/miscible with chloroform
- Pharmaceutical industry applications