Organic reference standard, codergocrine mesilate, Ph.Eur., RS
Catalog # EDQMC2510000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:8067-24-1
- Description:Codergocrine mesilate
- Formula:C₂₀H₂₉N₃O₅S
- Storage temperature:Ambient
- Description:Codergocrine mesilate
Specifications
About this item
- In Monographs 1416 and 2060