Organic reference standard, clozapine, Ph.Eur., RS
Catalog # EDQMC2460000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:5786-21-0
- Description:Clozapine
- Formula:C₁₈H₁₉CIN₄
- Storage temperature:Ambient
- Description:Clozapine
Specifications
About this item
- In Monograph 1191
- Water solubility: less than 1 mg/l
- Alcohol solubility: less than 1 mg/l