Organic reference standard, clotrimazole, Ph.Eur., RS
Catalog # EDQMC2430000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:23593-75-1
- Description:Clotrimazole
- Formula:C₂₂H₁₇ClN₂
- Storage temperature:Ambient
- Description:Clotrimazole
Specifications
About this item
- In Monograph 0757
- Soluble in/miscible with acetone, chloroform, ether, methanol