Order Entry
Singapore
ContactUsLinkComponent
Organic reference standards, Clomipramine, Ph.Eur., RS
Organic reference standards, Clomipramine, Ph.Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, Clomipramine, Ph.Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    17321-77-6
  • Formula:
    C₁₉H₂₄Cl₂N₂
  • Storage temperature:
    Ambient
  • Description:
    Clomipramine hydrochloride

Specifications