Organic reference standard, clenbuterol hydrochloride, Ph.Eur., RS
Catalog # EDQMC2248000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:30 mg
- Standards:Reference standard
- CAS No.:21898-19-1
- Description:Clenbuterol hydrochloride
- Formula:C₁₂H₁₉Cl₃N₂O
- Storage temperature:Ambient
- Description:Clenbuterol hydrochloride
Specifications
About this item
For use in the pharmaceutical industry.
- In Monograph 1409
- Soluble in water, DMSO, ethanol, and methanol