Organic reference standard, cinchocaine hydrochloride, Ph.Eur., RS
EDQMC2177000
: EDQM
:
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:61-12-1
- Description:Cinchocaine hydrochloride
- Formula:C₂₀H₃₀ClN₃O₂
- Storage temperature:Ambient
- Description:Cinchocaine hydrochloride
- Soluble in water
- Pharmaceutical industry applications