Organic reference standard, cimetidine hydrochloride, Ph.Eur., RS
EDQMC2175500
: EDQM
:
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:70059-30-2
- Description:Cimetidine hydrochloride
- Formula:C₁₀H₁₇ClN₆S
- Storage temperature:Ambient
- Description:Cimetidine hydrochloride
- Soluble in water
- Pharmaceutical industry applications