Order Entry
Singapore
ContactUsLinkComponent
Organic reference standard, cimetidine, Ph.Eur., RS
Organic reference standard, cimetidine, Ph.Eur., RS
Catalog # EDQMC2175000
Supplier:  EDQM
CAS Number:  
Organic reference standard, cimetidine, Ph.Eur., RS
Catalog # EDQMC2175000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    51481-61-9
  • Description:
    Cimetidine
  • Formula:
    C₁₀H₁₆N₆S
  • Storage temperature:
    Ambient
  • Description:
    Cimetidine

Specifications

About this item

  • Soluble/miscible in water, DMSO, and methanol