Organic reference standard, Cilazapril, Ph.Eur., RS
Catalog # EDQMC2174000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:50 mg
- Standards:Reference standard
- CAS No.:92077-78-6
- Description:Cilazapril
- Formula:C₂₂H₃₃N₃O₆
- Storage temperature:Ambient
- Description:Cilazapril
Specifications
About this item
- In Monograph 1499
- Solubility in water at 20°C: more than 0.001 g/l