Organic reference standard, Chlortalidone, Ph.Eur., RS
Catalog # EDQMC1950000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:77-36-1
- Description:Chlortalidone
- Formula:C₁₄H₁₁ClN₂O₄S
- Storage temperature:Ambient
- Description:Chlortalidone
Specifications
About this item
- Pharmaceutical industry applications