Organic reference standard, Chlorpromazine hydrochloride, Cargo Aircraft Only, Ph.Eur., RS
Catalog # EDQMC1900000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 mg
- Standards:Reference standard
- CAS No.:69-09-0
- Description:Chlorpromazine hydrochloride, Cargo Aircraft Only
- Formula:C₁₇H₂₀Cl₂N₂S
- Storage temperature:Ambient
- Description:Chlorpromazine hydrochloride, Cargo Aircraft Only
Specifications
About this item
- In Monograph 0475
- Water-soluble