Organic reference standard, Chlorothiazide, Ph.Eur., RS
EDQMC1700000
: EDQM
:
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:58-94-6
- Description:Chlorothiazide
- Formula:C₇H₆ClN₃O₄S₂
- Storage temperature:Ambient
- Description:Chlorothiazide
- Solubility in water at 30 °C: 266 g/l
- Pharmaceutical industry applications