Organic reference standard, Cephaeline hydrochloride, Ph.Eur., RS
EDQMC0700000
: EDQM
:
- Pack type:Vial
- Pk:15 mg
- Standards:Reference standard
- CAS No.:5853-29-2
- Description:Cephaeline hydrochloride
- Formula:C₂₈H₄₀Cl₂N₂O₄
- Storage temperature:Ambient
- Description:Cephaeline hydrochloride
- In Monographs 0081, 0093, 0094, 1530, and 1875