Organic reference standard, Cefoxitin sodium, Ph.Eur., RS
Catalog # EDQMC0688000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:33564-30-6
- Description:Cefoxitin sodium
- Formula:C₁₆H₁₆N₃NaO₇S₂
- Storage temperature:Ambient
- Description:Cefoxitin sodium
Specifications
About this item
- Pharmaceutical industry applications