Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standards, Carisoprodol, Ph.Eur., RS
Catalog # EDQMY0000035
Supplier:  EDQM
CAS Number:  
Organic reference standards, Carisoprodol, Ph.Eur., RS
Catalog # EDQMY0000035
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    78-44-4
  • Description:
    Carisoprodol
  • Formula:
    C₁₂H₂₄N₂O₄
  • Storage temperature:
    Ambient
  • Description:
    Carisoprodol

Specifications

About this item

  • In Monograph 1689
  • Solubility in water: 0,1 - 1 g/l