Organic reference standard, Bifonazole, Ph.Eur., RS
Catalog # EDQMB1110000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:60628-96-8
- Description:Bifonazole
- Formula:C₂₂H₁₈N₂
- Storage temperature:Ambient
- Description:Bifonazole
Specifications
About this item
- In Monograph 1395
- Solubility in water: less than 0,1 g/l