Organic reference standard, Bezafibrate, Ph.Eur., RS
Catalog # EDQMB1115000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:41859-67-0
- Description:Bezafibrate
- Formula:C₁₉H₂₀CINO₄
- Storage temperature:Ambient
- Description:Bezafibrate
Specifications
About this item
- In Monograph 1394