Organic reference standard, Betamethasone acetate, Ph.Eur., RS
EDQMB1030000
: EDQM
:
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:987-24-6
- Description:Betamethasone acetate
- Formula:C₂₄H₃₁FO₆
- Storage temperature:Ambient
- Description:Betamethasone acetate
- In Monographs 0548 and 0975
- Solubility in water: less than 0,1 g/l