Organic reference standards, Benserazide hydrochloride, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- In Monograph 1173
Specifications
- CAS No.:14919-77-8
- Formula:C₁₀H₁₆ClN₃O₅
- Storage temperature:Ambient
- Description:Benserazide hydrochloride