Organic reference standard, Benperidol, Ph.Eur., RS
Catalog # EDQMB0470000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 mg
- Standards:Reference standard
- CAS No.:2062-84-2
- Description:Benperidol
- Formula:C₂₂H₂₄FN₃O₂
- Storage temperature:+5°C ± 3°C
- Description:Benperidol
Specifications
About this item
- Method: Monograph only
- API product type
- Pharmaceutical industry applications