Organic reference standard, Anhydrotetracycline hydrochloride, Ph.Eur., RS
EDQMA1200000
: EDQM
:
- Pack type:Vial
- Pk:15 mg
- Standards:Reference standard
- CAS No.:13803-65-1
- Description:Anhydrotetracycline hydrochloride
- Formula:C₂₂H₂₃ClN₂O₇
- Storage temperature:Ambient
- Description:Anhydrotetracycline hydrochloride
- Method: Monograph only
- API product type
- Pharmaceutical industry applications