Organic reference standard, Amikacin, Ph.Eur., RS
Catalog # EDQMA0368000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 mg
- Standards:Reference standard
- CAS No.:1257517-67-1
- Description:Amikacin
- Formula:C₂₂H₄₅N₅O₁₄
- Description:Amikacin
Specifications
About this item
- Method: Monograph only
- API product type
- Pharmaceutical industry applications