Organic reference standard, 4-Epitetracycline hydrochloride, Ph.Eur., RS
EDQME0600000
: EDQM
:
- Pack type:Vial
- Pk:30 mg
- Standards:Reference standard
- CAS No.:23313-80-6
- Description:4-Epitetracycline hydrochloride
- Formula:C₂₂H₂₅ClN₂O₈
- Storage temperature:Ambient
- Description:4-Epitetracycline hydrochloride
- In Monographs 0210, 0211, and 1654