Organic reference standards, tolnaftates, Ph. Eur., RS
Catalog # EDQMT1707000
Supplier: EDQM
CAS Number:
Specifications
- Pk:60 mg
- Standards:Reference standard
- CAS No.:2398-96-1
- Description:Tolnaftate
- Seq:01
- Formula:C₁₉H₁₇NOS
- Description:Tolnaftate