Order Entry
Singapore
Orders LinkContactUsLinkComponent
Ubbelohde viscometers, calibrated
Ubbelohde viscometers, calibrated
Catalog #
Ubbelohde viscometers, calibrated
Catalog #
Supplier:  
Ubbelohde viscometers, calibrated
Product Details & Documents
These Ubbelohde viscometers are used to measure kinematic viscosity of transparent liquids.
  • Standard in petroleum industry
  • Well suited for use at very high and low temperatures
  • Viscometer constant does not vary with bath temperature
They suspend through a 510 mm hole of a constant temperature bath by means of a round holder and require a minimum sample of 11 ml and liquid bath depth of 241 mm.