Order Entry
Singapore
ContactUsLinkComponent
Crucibles, quartz
  TEGPFQVWR50CRU
Crucibles, quartz
  TEGPFQVWR50CRU
  • Ø×H:
    48×50 mm
  • Capacity:
    50 ml
  • Seq:
    02
  • Pk:
    1
These fused quartz crucibles and lids consists of high-purity silica (SiO₂) with a purity level of 99,998%, making them valuable in research and industry.
  • Optical transparency over a wide range
  • Low thermal expansion
  • Excellent thermal shock
  • Resistance to large numbers of elements and compounds
These crucibles offer several unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These properties include high purity, high softening temperature (1670 °C), optical transparency over a wide range.
 : Lids are sold separately.
Lids