Organic reference standards, nimesulide, Ph. Eur. RS
Catalog # EDQMN0845000
Supplier: EDQM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standard
- CAS No.:51803-78-2
- Description:Nimesulide
- Seq:01
- Formula:C₁₃H₁₂N₂O₅S
- Description:Nimesulide