Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standards, midazolam, Ph. Eur. RS
 :  EDQM
 
Organic reference standards, midazolam, Ph. Eur. RS
 :  EDQM

 

  • CAS No.:
    59468-44-9
  • Formula:
    C₁₉H₁₃ClFN₃O₂
  • Description:
    Midazolam impurity C, 8-chloro-6-(2-fluorophenyl)-1-methyl-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylic acid