Organic Reference Standards, Methyldopa, Ph. Eur. RS
Catalog # EDQMM1500000
Supplier: EDQM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standard
- CAS No.:41372-08-1
- Description:Methyldopa
- Seq:01
- Formula:C₁₀H₁₃NO₄
- Description:Methyldopa