Organic reference standards, lactuloses, Ph. Eur.
EDQMY0001510
: EDQM
:
- Pk:2.600 mg
- Standards:Reference standard
- CAS No.:4618-18-2
- Description:Lactulose for peak identification
- Seq:02
- Formula:C₁₂H₂₂O₁₁
- Description:Lactulose for peak identification