Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Acetate buffer solution pH 4.0, BDH®

4.0, ±0.05, Acetate, 1 L
Catalog # BDHB7300-1
Supplier:   BDH US PRODUCED
Acetate buffer solution pH 4.0, BDH®
Catalog # BDHB7300-1
Supplier:  BDH US PRODUCED
CAS Number:  
Acetate buffer solution pH 4.0, BDH®

4.0, ±0.05, Acetate, 1 L

CAS Number:  
Price
$145.00
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • pH buffer type:
    Acetate
  • Pk:
    1 L
  • pH accuracy:
    ±0.05
  • pH value:
    4.0

Specifications