Order Entry
Singapore
ContactUsLinkComponent
Silver nitrate 0.1 N
Silver nitrate 0.1 N
Catalog # BDHB7214-1
Supplier:  BDH US PRODUCED
CAS Number:  
undefined
Silver nitrate 0.1 N
Catalog # BDHB7214-1
Supplier:  BDH US PRODUCED
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Traceable to NIST SRM 919:
    Confirmed
  • Normality:
    0,0995 - 0,1005 N
  • Shelf-life:
    2 years

Specification Test Results

About this item

 :
 :169.87 g/mol
UN:3082
ADR:9,III
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414