Order Entry
Singapore
ContactUsLinkComponent
 
 
 
 
Sulphuric acid 95.0-98.0%, AR® ACS, Macron Fine Chemicals™
 
undefined
 
 
 
Sulphuric acid 95.0-98.0%, AR® ACS, Macron Fine Chemicals™
 

 

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance:
    Passes Test
  • ACS - Assay (H₂SO₄):
    95.0 - 98.0 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Ignition:
    ≤5 ppm
  • ACS - Chloride (Cl):
    ≤0.2 ppm
  • ACS - Nitrate (NO₃):
    ≤0.5 ppm
  • ACS - Ammonium (NH₄):
    ≤2 ppm
  • ACS - Substances Reducing Permanganate (as SO₂):
    ≤2 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Mercury (Hg):
    ≤5.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤0.010 ppm
  • Trace Impurities - Boron (B):
    ≤0.010 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.2 ppm