Order Entry
Singapore
ContactUsLinkComponent
 
Organic reference standards, econazoles, Ph. Eur.
  EDQMY0001236
 :  EDQM
 :  
Organic reference standards, econazoles, Ph. Eur.
  EDQMY0001236
 :  EDQM
 :  

 

  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    27220-47-9
  • Description:
    Econazole, Ph. Eur. standard
  • Seq:
    01
  • Formula:
    C₁₈H₁₅Cl₃N₂O
  • Description:
    Econazole