105° angled tube adapter with 24/40 Standard Taper joint, PYREX®
Catalog # CORN8840-24
Supplier: Corning
Specifications
- Socket size:24/40 NS
- Pk:12
Specifications
About this item
These angled tube adapters are made of PYREX® borosilicate glass.
- Arms are approximately 105° apart
- Upper end equipped with an outer 24/40 standard taper joint
- Lower end is cut off at an angle and is used as a receiver adapter
Tube Ext.Ø.: 12 mm