Order Entry
Singapore
ContactUsLinkComponent
75° angled tube adapter with two-way 24/40 Standard Taper joints, PYREX®
75° angled tube adapter with two-way 24/40 Standard Taper joints, PYREX®
Catalog # CORN8920-24
Supplier:  Corning
undefined
75° angled tube adapter with two-way 24/40 Standard Taper joints, PYREX®
Catalog # CORN8920-24
Supplier:  Corning
Specifications
  • Cone size:
    24/40 NS
  • Seq:
    01
  • Pk:
    12
Specifications
About this item
These angled two-way adapters are made of PYREX® borosilicate glass.
  • Arms are approximately 75° apart
  • Both ends are equipped with an inner 24/40 standard taper joint
It is often used as a stillhead adapter.