Order Entry
Singapore
ContactUsLinkComponent
105° angled tube adapter with 24/40 Standard Taper joint, PYREX®
105° angled tube adapter with 24/40 Standard Taper joint, PYREX®
Catalog # CORN8940-24
Supplier:  Corning
undefined
105° angled tube adapter with 24/40 Standard Taper joint, PYREX®
Catalog # CORN8940-24
Supplier:  Corning
Specifications
  • Cone size:
    24/40 NS
  • Socket size:
    24/40 NS
  • Pk:
    12
Specifications
About this item
These angled two-way adapters are made of PYREX® borosilicate glass.
  • Arms are approximately 105° apart
  • Used as a receiver adapter
  • Very good chemical and temperature resistance
One end is equipped with an outer 24/40 standard taper joint and the other end has an inner 24/40 standard taper joint. Used as a receiver adapter.