Volumetric pipettes, class A, color-coded with colored graduations, PYREX®
Supplier: Corning
Certificates
About this item
These precision PYREX® volumetric pipettes are manufactured and calibrated 'To Contain' (TC: bottom line) and 'To Deliver' (TD: top line) to specifications for Class A volumetric ware.
- Reusable
- Colour-coded with coloured graduations and calibration lines enameled onto the glass