Replacement cover for gum bomb, PYREX®
CORN7732-CO
: Corning
- Description:Cover for gum bomb
- Seq:901
- Pk:1
Replacement covers for gum bomb No. 7732. The cover prevents condensed vapors in bomb stem from contaminating the sample, but does not interfere with free flow of oxygen.
Made in accordance with ASTM D-525.