Order Entry
Singapore
ContactUsLinkComponent
Ethanol denatured (untaxed) (denatured with 1% MEK)
Irritant
Health
Flammable
Ethanol denatured (untaxed) (denatured with 1% MEK)
Catalog # HONC458600-25L
CAS Number:  
Irritant
Health
Flammable
Ethanol denatured (untaxed) (denatured with 1% MEK)
Catalog # HONC458600-25L
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Assay of Alcohol (from density):
    99.1 - 99.5 Vol.%
  • Non-volatile matter:
    Max. 0.010 %
  • Free acid (as CH₃COOH):
    Max. 0.003 %
  • APHA:
    max. 20
  • Refractive Index:
    n20/D 1.3600 (lit.)
  • Autoignition Temperature:
    683 °F
  • Vapor Density:
    1.59 (vs air)
  • Autoignition temp.:
    361,367 °C
  • Explosion limit:
    19%, 15,5 °C
  • Vapor pressure:
    44,6 mmHg ( 20 °C)
  • Freezing point:
    −115 °C

Specification Test Results

About this item

 :
 :46.07 g/mol
Boiling Pt:78…80 °C
Melting Pt:–144 °C
Density:0,78 g/cm3
Flash Pt:14 °C
UN:1987
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568