Order Entry
Singapore
Orders LinkContactUsLinkComponent
 

Sedimentation cones, Imhoff type, PYREX®

Imhoff cones, O. Ø 110 mm, height 435 mm
  CORN2180-439
 :   Corning
Sedimentation cones, Imhoff type, PYREX®
  CORN2180-439
 :  Corning
Sedimentation cones, Imhoff type, PYREX®
Sedimentation cones, Imhoff type, PYREX®

Imhoff cones, O. Ø 110 mm, height 435 mm

 
$2,000.00
   
1 * 4 items
 

Made of PYREX® borosilicate glass.

  • Also graduated and marked at 1000 ml
  • Blunt tip increases ruggedness and cleaning ease
  • Very good chemical and temperature resistance

These blunt tip cones are used with heavy sediments, where ability to read small volumes is unimportant. Graduated from 0 ml to 1 ml in 0,1 ml increments, 1 to 10 ml in 0,5 ml increments and 10 ml to 40 ml in 1 ml increments.

 

 
  • Description:
    Imhoff cones, O. Ø 110 mm, height 435 mm